Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172643.5 | NP_766231.2 | 8 | 1951 | ||
GenBank mRNA | AK034756.1 | - | 7 | 1048 | ||
AK036544.1 | - | 8 | 1950 | |||
AK079719.1 | - | 4 | 309 | |||
AK163302.1 | - | 7 | 1224 | |||
BC046465.1 | - | 7 | 1120 | |||
BC150859.1 | - | 7 | 1290 | 9087770 | ||
BC172012.1 | - | 8 | 1216 |