siRNA Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 6 | 1090 | ||
GenBank mRNA | AK030952.1 | - | 6 | 1079 | ||
AK033634.1 | - | 5 | 1913 | |||
AK037337.1 | - | 6 | 1109 | |||
AK048454.1 | - | 6 | 1067 | |||
AK138809.1 | - | 6 | 1091 | |||
AK154133.1 | - | 6 | 1119 | |||
AK154348.1 | - | 6 | 1119 | |||
AK164885.1 | - | 6 | 1060 | |||
BC057943.1 | - | 6 | 1114 | |||
BC076606.1 | - | 6 | 1082 | 30625515 | ||
BC089191.1 | - | 6 | 1064 | 30473852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
