siRNA Details
Gene Symbol: Lyplal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146106.2 | NP_666218.2 | 2 | 201 | ||
GenBank mRNA | AK148533.1 | - | 2 | 169 | ||
BC027340.1 | - | 2 | 201 | 4023059 |

Gene Symbol: Lyplal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146106.2 | NP_666218.2 | 2 | 201 | ||
GenBank mRNA | AK148533.1 | - | 2 | 169 | ||
BC027340.1 | - | 2 | 201 | 4023059 |