siRNA Details
Gene Symbol: Lyplal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146106.2 | NP_666218.2 | 3 | 335 | ||
GenBank mRNA | AK148533.1 | - | 3 | 303 | ||
BC027340.1 | - | 3 | 335 | 4023059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
