Search
Search

Hhat| Gene Symbol: Hhat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144881.4 | NP_659130.2 | 5 | 568 | ||
| GenBank mRNA | AK028230.1 | - | 5 | 371 | ||
| AK134661.1 | - | 5 | 411 | |||
| AK172126.1 | - | 5 | 415 | |||
| BC008159.1 | - | 562 | 3963716 | |||