Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033289.2 | NP_001028461.2 | 9 | 1978 | ||
GenBank mRNA | AF139193.1 | - | 4 | 331 | ||
AK077026.1 | - | 9 | 2070 | |||
AK138779.1 | - | 9 | 2355 | |||
BC103685.1 | - | 9 | 1978 | 40048593 | ||
BC104737.1 | - | 9 | 1978 | 40048591 | ||
BC105676.1 | - | 9 | 1977 |