Search
Search

Exosc2| Gene Symbol: Exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144886.2 | NP_659135.1 | 8 | 772 | ||
| GenBank mRNA | AK144234.1 | - | 8 | 781 | ||
| AK164618.1 | - | 7 | 1554 | |||
| AK166881.1 | - | 8 | 782 | |||
| BC021485.1 | - | 8 | 796 | 5344620 | ||
| BC021807.1 | - | 8 | 794 | 3980155 | ||