Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040689.1 | NP_001035779.1 | 1 | 210 | ||
GenBank mRNA | AK020904.1 | - | 1 | 211 | ||
AK043484.1 | - | 1 | 211 | |||
BC048707.1 | - | 1 | 184 | 6511472 | ||
BC116367.1 | - | 1 | 173 |