Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213733.2 | NP_998898.1 | 5 | 718 | ||
GenBank mRNA | AK144651.1 | - | 5 | 691 | ||
AK150313.1 | - | 5 | 719 | |||
AK156136.1 | - | 5 | 719 | |||
AK170509.1 | - | 5 | 719 | |||
AK178294.1 | - | 3 | 222 | |||
BC023239.1 | - | 4 | 396 | |||
BC055448.1 | - | 4 | 393 | |||
BC069933.1 | - | 5 | 713 | 4008091 | ||
BC080707.1 | - | 5 | 591 |