Search
Search

Clrn1| Gene Symbol: Clrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153384.3 | NP_700433.1 | 4 | 780 | ||
| NM_153385.3 | NP_700434.1 | 3 | 726 | |||
| NM_153386.3 | NP_700435.1 | 2 | 546 | |||
| GenBank mRNA | AF495718.1 | - | 4 | 780 | ||
| AF495719.1 | - | 3 | 726 | |||
| AF495720.1 | - | 2 | 544 | |||
| AK037267.1 | - | 4 | 444 | |||
| AK041623.1 | - | 2 | 482 | |||
| AK042523.1 | - | 2 | 482 | |||
| BC132254.1 | - | 2 | 528 | 40130531 | ||
| BC132256.1 | - | 2 | 528 | 40130533 | ||
| BC145202.1 | - | 3 | 708 | 9053753 | ||