Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165949.1 | NP_001159421.1 | 10 | 1318 | ||
NM_018812.2 | NP_061282.2 | 11 | 1213 | |||
NM_146135.2 | NP_666247.1 | 10 | 1355 | |||
GenBank mRNA | AF034080.3 | - | 10 | 1090 | ||
AK040619.1 | - | 10 | 1346 | |||
AK077551.1 | - | 10 | 1304 | |||
AK085503.1 | - | 10 | 1290 | |||
AK134404.1 | - | 5 | 493 | |||
BC023128.1 | - | 10 | 1261 | 3585764 | ||
BC050988.1 | - | 11 | 1172 | |||
BC051252.1 | - | 10 | 1228 | 6489803 |