Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACNAT1 SIRNA Assays ›
Gene Symbol:
Acnat1
Gene Name:
acyl-coenzyme A amino acid N-acyltransferase 1
Gene Aliases:
AI132189
Chromosome Location:
Chr.4: 49443532 - 49481312 on Build GRCm38
UniGene ID:
Mm.30101
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s106417
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acnat1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164565.1
NP_001158037.1
3
799
GenBank mRNA
DQ469311.1
-
3
799
Back To Top
More Information
Panther Classification:
Molecular Function -
esterase
hydrolase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
thiolester hydrolase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0