Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163470.1 | NP_001156942.1 | 10 | 1454 | ||
NM_172275.2 | NP_758479.2 | 10 | 1454 | |||
GenBank mRNA | AK042299.1 | - | 2 | 98 | ||
AK150038.1 | - | 10 | 1423 | |||
BC023072.1 | - | 4 | 1858 | |||
BC038396.1 | - | 10 | 1410 | 3987227 |