Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146176.3 | NP_666288.1 | 6, 7 | 760 | ||
GenBank mRNA | AK141718.1 | - | 6, 7 | 740 | ||
AK173003.1 | - | 2, 3 | 174 | |||
BC030332.1 | - | 6, 7 | 689 | 5400871 | ||
BC053437.1 | - | 6, 7 | 521 | 6394085 | ||
BC141041.1 | - | 6, 7 | 560 | 9055653 |