Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PMS2 SIRNA Assays ›
          Gene Symbol:
          PMS2 PMS2CL
          Gene Name:
          PMS1 homolog 2, mismatch repair system component
          PMS2 C-terminal like pseudogene
          Gene Aliases:
          HNPCC4, MLH4, PMS2CL, PMSL2
          PMS2P13
          Chromosome Location:
          Chr.7: 5970925 - 6009106 on Build GRCh38
          Chromosome Location:
          Chr.7: 6735305 - 6751601 on Build GRCh38
          UniGene ID:
          Hs.632637
          Hs.715590
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s10741

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PMS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000535.6 NP_000526.2 11 2070
          NM_001322003.1 NP_001308932.1 11 2065
          NM_001322004.1 NP_001308933.1 10 1930
          NM_001322005.1 NP_001308934.1 11 2182
          NM_001322006.1 NP_001308935.1 10 1914
          NM_001322007.1 NP_001308936.1 10 1967
          NM_001322008.1 NP_001308937.1 9 1827
          NM_001322009.1 NP_001308938.1 11 2177
          NM_001322010.1 NP_001308939.1 9 1774
          NM_001322011.1 NP_001308940.1 11 2021
          NM_001322012.1 NP_001308941.1 11 2016
          NM_001322013.1 NP_001308942.1 10 1897
          NM_001322014.1 NP_001308943.1 11 2070
          NM_001322015.1 NP_001308944.1 11 2240
          NR_136154.1 11 2070
          XM_006715744.3 XP_006715807.1 4 1146
          XM_017012342.1 XP_016867831.1 10 1895
          GenBank mRNA AB103086.1 - 8 1665
          AK294661.1 - 10 1920
          AK312390.1 - 11 2019
          BC031832.1 - 11 2034
          BC093921.1 - 11 2005 7939766
          U14658.1 - 11 1985
          Gene Symbol: PMS2CL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002217.1 2 923
          GenBank mRNA AB116525.1 - 2 923
          BC041364.1 - 9 1678
          CR749616.1 - 6 1262

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mismatch repair
          somatic hypermutation of immunoglobulin genes
          response to drug
          nucleic acid phosphodiester bond hydrolysis
          DNA binding
          single-stranded DNA binding
          endonuclease activity
          protein binding
          ATP binding
          ATPase activity
          single base insertion or deletion binding
          MutSalpha complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0