Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT4 SIRNA Assays ›
Gene Symbol:
SEPT4
Gene Name:
septin 4
Gene Aliases:
ARTS, BRADEION, CE5B3, H5, MART, PNUTL2, SEP4, hCDCREL-2, hucep-7
Chromosome Location:
Chr.17: 58520250 - 58540818 on Build GRCh38
UniGene ID:
Hs.287518
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10767
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198713.1
NP_001185642.1
11
1447
NM_001256782.1
NP_001243711.1
12
1544
NM_001256822.1
NP_001243751.1
11
1516
NM_004574.4
NP_004565.1
11
1647
NM_080415.3
NP_536340.1
9
1686
NM_080416.3
NP_536341.1
11
1418
NR_037155.2
9
2170
NR_104196.1
10
1802
NR_104197.1
10
1541
XM_006721949.2
XP_006722012.1
12
1476
XM_006721950.3
XP_006722013.1
11
1168
XM_006721951.2
XP_006722014.1
10
3302
XM_006721952.2
XP_006722015.1
10
1121
XM_006721954.2
XP_006722017.1
11
1161
XM_006721955.2
XP_006722018.1
11
3344
XM_011524911.1
XP_011523213.1
10
1075
XM_011524912.1
XP_011523214.1
11
1833
GenBank mRNA
AB002110.1
-
9
1973
AB008753.1
-
11
1482
AF035811.1
-
11
1483
AF073312.1
-
11
1478
AF176379.1
-
9
1652
AK091065.1
-
6
1944
AK094579.1
-
10
2539
AK294094.1
-
11
1450
AK301914.1
-
12
1544
AK302146.1
-
11
1447
AK315045.1
-
10
1649
AK315396.1
-
11
1372
BC018055.1
-
10
1458
BC018056.1
-
11
1361
4811662
CR457111.1
-
11
1354
D89278.1
-
11
1354
U88829.1
-
11
1479
U88870.1
-
11
1364
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
cell cycle
brain development
sperm mitochondrion organization
positive regulation of protein ubiquitination
regulation of apoptotic process
positive regulation of apoptotic process
sperm capacitation
cell division
positive regulation of intrinsic apoptotic signaling pathway
GTPase activity
structural molecule activity
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0