Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEPT4 SIRNA Assays ›
          Gene Symbol:
          SEPT4
          Gene Name:
          septin 4
          Gene Aliases:
          ARTS, BRADEION, CE5B3, H5, MART, PNUTL2, SEP4, hCDCREL-2, hucep-7
          Chromosome Location:
          Chr.17: 58520250 - 58540818 on Build GRCh38
          UniGene ID:
          Hs.287518
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10768

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEPT4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198713.1 NP_001185642.1 8, 9 1103
          NM_001256782.1 NP_001243711.1 9, 10 1200
          NM_001256822.1 NP_001243751.1 8, 9 1172
          NM_004574.4 NP_004565.1 8, 9 1303
          NM_080415.3 NP_536340.1 7, 8 1187
          NM_080416.3 NP_536341.1 8, 9 1074
          NR_037155.2 7, 8 1671
          NR_104196.1 8, 9 1303
          NR_104197.1 7, 8 1197
          XM_006721949.2 XP_006722012.1 9, 10 1132
          XM_006721950.3 XP_006722013.1 8, 9 824
          XM_006721951.2 XP_006722014.1 7, 8 2958
          XM_006721952.2 XP_006722015.1 7, 8 777
          XM_006721954.2 XP_006722017.1 8, 9 817
          XM_006721955.2 XP_006722018.1 8, 9 3000
          XM_011524911.1 XP_011523213.1 7, 8 731
          XM_011524912.1 XP_011523214.1 8, 9 1489
          GenBank mRNA AB002110.1 - 7, 8 1474
          AB008753.1 - 8, 9 1138
          AF035811.1 - 8, 9 1139
          AF073312.1 - 8, 9 1134
          AF176379.1 - 7, 8 1153
          AK094579.1 - 7, 8 2195
          AK294094.1 - 8, 9 1106
          AK301914.1 - 9, 10 1200
          AK302146.1 - 8, 9 1103
          AK315045.1 - 8, 9 1150
          AK315396.1 - 8, 9 1028
          BC018055.1 - 7, 8 1114
          BC018056.1 - 8, 9 1017 4811662
          CR457111.1 - 8, 9 1010
          D89278.1 - 8, 9 1010
          U88829.1 - 8, 9 1135
          U88870.1 - 8, 9 1020

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001011893, XM_008767966, AY208293, BC088334, AY208294, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011129, NM_001284392, NM_001284394, NM_001284398, BC055101, and others.

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          cell cycle
          brain development
          sperm mitochondrion organization
          positive regulation of protein ubiquitination
          regulation of apoptotic process
          positive regulation of apoptotic process
          sperm capacitation
          cell division
          positive regulation of intrinsic apoptotic signaling pathway
          GTPase activity
          structural molecule activity
          protein binding
          GTP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0