Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145605.2 | NP_663580.2 | 10, 11 | 1528 | ||
GenBank mRNA | AK089954.1 | - | 10, 11 | 1492 | ||
AK161674.1 | - | 10, 11 | 1504 | |||
BC012312.1 | - | 10, 11 | 1528 | 2631632 | ||
BC023738.1 | - | 10, 11 | 1499 | 5352770 |