Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172769.2 | NP_766357.1 | 6 | 888 | ||
GenBank mRNA | AB016248.1 | - | 6 | 874 | ||
AK043825.1 | - | 6 | 847 | |||
AK077670.1 | - | 6 | 846 | |||
AK154309.1 | - | 6 | 835 | |||
BC024132.1 | - | 6 | 837 | 5101986 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK052921.1 | - | 749 | Chr. - - [-] on Build |