Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADORA1 SIRNA Assays ›
          Gene Symbol:
          ADORA1
          Gene Name:
          adenosine A1 receptor
          Gene Aliases:
          RDC7
          Chromosome Location:
          Chr.1: 203127705 - 203167405 on Build GRCh38
          UniGene ID:
          Hs.77867
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1086

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADORA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000674.2 NP_000665.1 4 1049
          NM_001048230.1 NP_001041695.1 3 894
          XM_005244901.1 XP_005244958.1 4 1257
          XM_005244902.4 XP_005244959.1 2 824
          XM_017000342.1 XP_016855831.1 3 1056
          GenBank mRNA AB004662.1 - 2 616
          AK095060.1 - 4 842
          AK127752.1 - 3 1537
          AK289928.1 - 4 1049
          AK293647.1 - 3 982
          AK295584.1 - 3 719
          AK308733.1 - 4 866
          AK316106.1 - 3 975
          AY136746.1 - 2 616
          BC026340.1 - 3 872 4797116
          BT019854.1 - 2 616
          CR541749.1 - 2 616
          EF057066.1 - 2 821
          KC881108.1 - 2 434
          KC884744.1 - 2 335
          L22214.1 - 6 1026
          S45235.1 - 3 700
          S56143.1 - 2 682
          X68485.1 - 3 1053

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          temperature homeostasis
          response to hypoxia
          adenosine receptor signaling pathway
          regulation of respiratory gaseous exchange by neurological system process
          negative regulation of acute inflammatory response
          negative regulation of leukocyte migration
          positive regulation of peptide secretion
          regulation of glomerular filtration
          protein targeting to membrane
          phagocytosis
          inflammatory response
          signal transduction
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          cell-cell signaling
          nervous system development
          negative regulation of cell proliferation
          negative regulation of glutamate secretion
          lipid catabolic process
          negative regulation of synaptic transmission, GABAergic
          positive regulation of nucleoside transport
          negative regulation of neurotrophin production
          positive regulation of protein dephosphorylation
          negative regulation of renal sodium excretion
          negative regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of epidermal growth factor-activated receptor activity
          negative regulation of blood pressure
          positive regulation of blood pressure
          negative regulation of vasodilation
          negative regulation of hormone secretion
          cognition
          detection of temperature stimulus involved in sensory perception of pain
          negative regulation of lipid catabolic process
          regulation of sensory perception of pain
          negative regulation of synaptic transmission, glutamatergic
          negative regulation of cardiac muscle contraction
          excitatory postsynaptic potential
          relaxation of vascular smooth muscle
          negative regulation of mucus secretion
          apoptotic signaling pathway
          negative regulation of long term synaptic depression
          G-protein coupled adenosine receptor activity
          G-protein coupled receptor binding
          purine nucleoside binding
          phospholipase C activity
          protein binding
          heat shock protein binding
          G-protein beta/gamma-subunit complex binding
          heterotrimeric G-protein binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tg9dg:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0