Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPM1A SIRNA Assays ›
Gene Symbol:
PPM1A
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases:
PP2C-ALPHA, PP2CA, PP2Calpha
Chromosome Location:
Chr.14: 60245752 - 60299087 on Build GRCh38
UniGene ID:
Hs.130036
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10920
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPM1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021003.4
NP_066283.1
2
1184
NM_177951.2
NP_808820.1
3
1286
NM_177952.2
NP_808821.2
2
1049
XM_005267779.1
XP_005267836.1
3
1131
XM_005267781.1
XP_005267838.1
3
1289
XM_011536872.1
XP_011535174.1
5
1551
XM_011536876.1
XP_011535178.1
4
1408
XM_011536877.1
XP_011535179.1
5
1451
XM_011536878.1
XP_011535180.1
4
1374
XM_011536879.2
XP_011535181.1
4
1243
XM_011536880.2
XP_011535182.1
5
1297
XM_011536881.2
XP_011535183.1
3
1157
XM_011536882.2
XP_011535184.1
2
826
XM_011536883.1
XP_011535185.1
3
1240
XM_011536884.1
XP_011535186.1
2
797
XM_017021381.1
XP_016876870.1
5
2421
XM_017021382.1
XP_016876871.1
7
1671
XM_017021383.1
XP_016876872.1
4
1307
XM_017021384.1
XP_016876873.1
4
999
XM_017021385.1
XP_016876874.1
3
1274
XM_017021386.1
XP_016876875.1
3
1037
XM_017021387.1
XP_016876876.1
4
1283
GenBank mRNA
AB451247.1
-
1
734
AB451371.1
-
1
734
AF070670.1
-
3
1093
AK097843.1
-
2
1049
AK308225.1
-
2
911
AK313337.1
-
3
1131
BC026691.1
-
2
1037
3847638
BC063243.1
-
2
967
6011486
S87759.1
-
2
1091
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00221372_m1
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein dephosphorylation
N-terminal protein myristoylation
cell cycle arrest
negative regulation of SMAD protein complex assembly
Wnt signaling pathway
dephosphorylation
positive regulation of Wnt signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of transcription, DNA-templated
positive regulation of protein export from nucleus
cellular response to transforming growth factor beta stimulus
magnesium ion binding
protein serine/threonine phosphatase activity
signal transducer activity
protein binding
manganese ion binding
calmodulin-dependent protein phosphatase activity
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0