Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPM1A SIRNA Assays ›
Gene Symbol:
PPM1A
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases:
PP2C-ALPHA, PP2CA, PP2Calpha
Chromosome Location:
Chr.14: 60245752 - 60299087 on Build GRCh38
UniGene ID:
Hs.130036
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10921
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPM1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021003.4
NP_066283.1
2
697
NM_177951.2
NP_808820.1
3
799
NM_177952.2
NP_808821.2
2
562
XM_005267779.1
XP_005267836.1
3
644
XM_005267781.1
XP_005267838.1
3
802
XM_011536872.1
XP_011535174.1
5
1064
XM_011536876.1
XP_011535178.1
4
921
XM_011536877.1
XP_011535179.1
5
964
XM_011536878.1
XP_011535180.1
4
887
XM_011536879.2
XP_011535181.1
4
756
XM_011536880.2
XP_011535182.1
5
810
XM_011536881.2
XP_011535183.1
3
670
XM_011536882.2
XP_011535184.1
2
339
XM_011536883.1
XP_011535185.1
3
753
XM_011536884.1
XP_011535186.1
2
310
XM_017021381.1
XP_016876870.1
5
1934
XM_017021382.1
XP_016876871.1
7
1184
XM_017021383.1
XP_016876872.1
4
820
XM_017021384.1
XP_016876873.1
4
512
XM_017021385.1
XP_016876874.1
3
787
XM_017021386.1
XP_016876875.1
3
550
XM_017021387.1
XP_016876876.1
4
796
GenBank mRNA
AB451247.1
-
1
247
AB451371.1
-
1
247
AF070670.1
-
3
606
AK097843.1
-
2
562
AK308225.1
-
2
426
AK313337.1
-
3
644
BC026691.1
-
2
550
3847638
BC063243.1
-
2
480
6011486
S87759.1
-
2
604
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_017038, XM_006240195, XM_006240194, XM_006240193, J04503.
Matching Taqman Gene Expression Assay:
Hs00221372_m1
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein dephosphorylation
N-terminal protein myristoylation
cell cycle arrest
negative regulation of SMAD protein complex assembly
Wnt signaling pathway
dephosphorylation
positive regulation of Wnt signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of transcription, DNA-templated
positive regulation of protein export from nucleus
cellular response to transforming growth factor beta stimulus
magnesium ion binding
protein serine/threonine phosphatase activity
signal transducer activity
protein binding
manganese ion binding
calmodulin-dependent protein phosphatase activity
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0