Search
Search

Adamts19| Gene Symbol: Adamts19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175506.3 | NP_780715.1 | 2 | 156 | ||
| GenBank mRNA | AK053213.1 | - | 2 | 528 | ||
| AK165144.1 | - | 1 | 143 | |||
| AY135183.1 | - | 2 | 156 | |||
| BC150736.1 | - | 2 | 253 | 9087647 | ||