siRNA Details
Gene Symbol: Ccdc108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039495.1 | NP_001034584.1 | 26, 27 | 4475 | ||
GenBank mRNA | AK133278.1 | - | 26, 27 | 4476 | ||
BC150939.1 | - | 26, 27 | 4421 | 9087850 |
