Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TECRL SIRNA Assays ›
          Gene Symbol:
          Tecrl
          Gene Name:
          trans-2,3-enoyl-CoA reductase-like
          Gene Aliases:
          D330017N19Rik, Srd5a2l2
          Chromosome Location:
          Chr.5: 83278122 - 83355195 on Build GRCm38
          UniGene ID:
          Mm.126783
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s109921

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tecrl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_153801.3 NP_722496.2 10 1088
          GenBank mRNA AF548365.1 - 10 1089
          AK052276.1 - 10 927
          AK052284.1 - 10 1091
          AK052302.1 - 10 793
          AK052378.1 - 10 851
          BC107366.1 - 10 1001 40056219
          BC107367.1 - 10 1001 40056223

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          biological_process
          oxidation-reduction process
          molecular_function
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-CH group of donors
          enoyl-[acyl-carrier-protein] reductase activity
          2,3-dihydroxy-2,3-dihydro-phenylpropionate dehydrogenase activity
          cis-2,3-dihydrodiol DDT dehydrogenase activity
          trans-9R,10R-dihydrodiolphenanthrene dehydrogenase activity
          cis-chlorobenzene dihydrodiol dehydrogenase activity
          2,5-dichloro-2,5-cyclohexadiene-1,4-diol dehydrogenase activity
          trans-1,2-dihydrodiolphenanthrene dehydrogenase activity
          3,4-dihydroxy-3,4-dihydrofluorene dehydrogenase activity
          benzo(a)pyrene-trans-11,12-dihydrodiol dehydrogenase activity
          benzo(a)pyrene-cis-4,5-dihydrodiol dehydrogenase activity
          citronellyl-CoA dehydrogenase activity
          menthone dehydrogenase activity
          phthalate 3,4-cis-dihydrodiol dehydrogenase activity
          cinnamate reductase activity
          NADPH-dependent curcumin reductase activity
          NADPH-dependent dihydrocurcumin reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0