Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP3CB SIRNA Assays ›
          Gene Symbol:
          PPP3CB
          Gene Name:
          protein phosphatase 3 catalytic subunit beta
          Gene Aliases:
          CALNA2, CALNB, CNA2, PP2Bbeta
          Chromosome Location:
          Chr.10: 73436428 - 73496024 on Build GRCh38
          UniGene ID:
          Hs.500067
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11004
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP3CB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142353.2 NP_001135825.1 2 320
          NM_001142354.2 NP_001135826.1 2 320
          NM_001289968.1 NP_001276897.1 2 320
          NM_001289969.1 NP_001276898.1 3 439
          NM_021132.3 NP_066955.1 2 320
          XM_005269944.1 XP_005270001.1 2 320
          XM_017016379.1 XP_016871868.1 2 320
          GenBank mRNA AJ488506.1 - 2 257
          AK301582.1 - 3 346
          BC028049.1 - 2 308 4538822
          BC035464.1 - 2 278
          DA854064.1 - 2 320
          M29550.1 - 2 292
          M29551.1 - 2 301

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00184176_m1

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of T cell mediated cytotoxicity
          lymphangiogenesis
          protein phosphorylation
          protein dephosphorylation
          signal transduction
          Wnt signaling pathway, calcium modulating pathway
          heart development
          learning
          memory
          dephosphorylation
          calcium ion regulated exocytosis
          T cell differentiation
          locomotion involved in locomotory behavior
          calcineurin-NFAT signaling cascade
          response to cytokine
          social behavior
          cellular response to drug
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          Fc-epsilon receptor signaling pathway
          T cell proliferation
          T cell activation
          T cell homeostasis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of synaptic plasticity
          axon extension
          regulation of insulin secretion
          positive regulation of NFAT protein import into nucleus
          protein serine/threonine phosphatase activity
          calcium ion binding
          protein binding
          calmodulin binding
          drug binding
          enzyme binding
          protein phosphatase 2B binding
          calmodulin-dependent protein phosphatase activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline