Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172893.3 | NP_766481.2 | 3 | 815 | ||
GenBank mRNA | AK036886.1 | - | 3 | 818 | ||
AK053205.1 | - | 3 | 715 | |||
AK081528.1 | - | 3 | 714 | |||
AK156623.1 | - | 3 | 684 | |||
AK162200.1 | - | 3 | 386 | |||
AK206233.1 | - | 2 | 257 | |||
BC120733.1 | - | 3 | 648 | 40129656 | ||
BC137645.1 | - | 3 | 648 | 8860664 |