Search
Search

Nlrp10| Gene Symbol: Nlrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175532.3 | NP_780741.1 | 3 | 1014 | ||
| GenBank mRNA | AK028575.1 | - | 3 | 1037 | ||
| AK028802.1 | - | 3 | 1016 | |||
| AK032446.1 | - | 3 | 1010 | |||
| AY355344.1 | - | 2 | 983 | |||
| AY489193.1 | - | 2 | 967 | |||
| BC113194.1 | - | 3 | 1010 | 40062457 | ||
| BC113801.1 | - | 3 | 1010 | 40062458 | ||