Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11069
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
11
934
NM_001242858.2
NP_001229787.1
8
877
NM_001242859.2
NP_001229788.1
10
925
NM_001242860.2
NP_001229789.1
11
1069
NM_001242861.2
NP_001229790.1
9
961
NM_001242862.2
NP_001229791.1
7
673
NM_001300915.1
NP_001287844.1
11
1069
NM_001300916.1
NP_001287845.1
8
939
NM_001300917.1
NP_001287846.1
8
877
NM_002731.3
NP_002722.1
8
1035
NM_182948.3
NP_891993.1
8
939
NM_207578.2
NP_997461.1
8
1035
XM_005271015.3
XP_005271072.1
10
886
XM_005271016.1
XP_005271073.1
10
1011
XM_005271017.3
XP_005271074.1
9
876
XM_005271018.4
XP_005271075.1
9
1196
XM_005271019.1
XP_005271076.1
8
952
XM_005271020.1
XP_005271077.1
8
919
XM_006710758.1
XP_006710821.1
10
1025
XM_017001706.1
XP_016857195.1
11
929
XM_017001707.1
XP_016857196.1
11
929
XM_017001708.1
XP_016857197.1
11
1030
XM_017001709.1
XP_016857198.1
10
727
XM_017001710.1
XP_016857199.1
10
1025
XM_017001711.1
XP_016857200.1
10
727
XM_017001712.1
XP_016857201.1
10
1025
XM_017001713.1
XP_016857202.1
8
977
XM_017001714.1
XP_016857203.1
10
1011
XM_017001715.1
XP_016857204.1
10
1011
XM_017001716.1
XP_016857205.1
9
879
XM_017001717.1
XP_016857206.1
9
1200
XM_017001718.1
XP_016857207.1
8
854
XM_017001719.1
XP_016857208.1
8
952
XM_017001720.1
XP_016857209.1
8
952
GenBank mRNA
AB209189.1
-
11
1005
AB451239.1
-
8
684
AB451364.1
-
8
684
AK091420.1
-
9
635
AK296482.1
-
11
736
AK304322.1
-
8
864
AK304375.1
-
8
679
AK314550.1
-
8
931
AK316115.1
-
7
646
BC016285.1
-
8
866
3916100
BC035058.1
-
8
953
5262372
BI598037.1
-
8
750
BM905428.1
-
8
823
BX369882.1
-
1
96
BX537705.1
-
8
916
BX641026.1
-
8
915
BX647515.1
-
11
1014
CR936631.1
-
10
728
M34181.1
-
8
731
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00176944_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0