Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144897.3 | NP_659146.1 | 6 | 695 | ||
GenBank mRNA | AJ344092.1 | - | 6 | 695 | ||
AK131615.1 | - | 6 | 906 | |||
AK159846.1 | - | 6 | 697 | |||
AY566271.1 | - | 6 | 695 | |||
BC050084.1 | - | 1 | 34 | |||
BC058362.1 | - | 6 | 693 | 6819459 |