Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF373288.1 | - | 1 | 61 | ||
AK029590.1 | - | 1 | 33 | |||
BC132102.1 | - | 1 | 26 | 40130379 | ||
BC132292.1 | - | 1 | 26 | 40130569 | ||
BC141203.1 | - | 1 | 26 | 9055815 | ||
BC141204.1 | - | 1 | 26 | 9055816 |
Gene Symbol: Gm5347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079931.2 | NP_001073400.1 | 1 | 30 |