Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK1 SIRNA Assays ›
          Gene Symbol:
          MAPK1
          Gene Name:
          mitogen-activated protein kinase 1
          Gene Aliases:
          ERK, ERK-2, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, p42-MAPK
          Chromosome Location:
          Chr.22: 21759657 - 21867680 on Build GRCh38
          UniGene ID:
          Hs.431850
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11139
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002745.4 NP_002736.3 5 903
          NM_138957.3 NP_620407.1 5 903
          GenBank mRNA AK090869.1 - 6 760
          AK295188.1 - 7 836
          AK307615.1 - 6 748
          BC017832.1 - 5 903 4685557
          BC099905.1 - 5 868 5264558
          EF565108.1 - 3 349
          Z11694.1 - 5 847
          Z11695.1 - 5 761

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00177066_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          MAPK import into nucleus
          transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          chemotaxis
          response to stress
          cellular response to DNA damage stimulus
          cell cycle
          signal transduction
          chemical synaptic transmission
          axon guidance
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          response to toxic substance
          positive regulation of peptidyl-threonine phosphorylation
          regulation of phosphatidylinositol 3-kinase signaling
          viral process
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          sensory perception of pain
          cytosine metabolic process
          platelet activation
          positive regulation of cell migration
          thyroid gland development
          regulation of protein stability
          lipopolysaccharide-mediated signaling pathway
          positive regulation of telomere maintenance via telomerase
          regulation of stress-activated MAPK cascade
          mammary gland epithelial cell proliferation
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB signaling pathway
          outer ear morphogenesis
          response to exogenous dsRNA
          response to estrogen
          negative regulation of cell differentiation
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          thymus development
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          regulation of sequence-specific DNA binding transcription factor activity
          regulation of cytoskeleton organization
          positive regulation of telomerase activity
          Bergmann glial cell differentiation
          long-term synaptic potentiation
          face development
          JAK-STAT cascade involved in growth hormone signaling pathway
          lung morphogenesis
          trachea formation
          labyrinthine layer blood vessel development
          cardiac neural crest cell development involved in heart development
          ERK1 and ERK2 cascade
          response to epidermal growth factor
          caveolin-mediated endocytosis
          regulation of Golgi inheritance
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          regulation of cellular response to heat
          positive regulation of telomere capping
          regulation of early endosome to late endosome transport
          phosphotyrosine binding
          DNA binding
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          transcription factor binding
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          phosphatase binding
          mitogen-activated protein kinase kinase kinase binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline