Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207135.2 | NP_997018.2 | 1 | 256 | ||
GenBank mRNA | CB173458.1 | - | 1 | 264 |
Gene Symbol: Olfr1080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146409.2 | NP_666521.2 | 2 | 281 | ||
GenBank mRNA | CB174209.1 | - | 2 | 281 |
Gene Symbol: Olfr1082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207674.2 | NP_997557.2 | 2 | 292 | ||
GenBank mRNA | CB173611.1 | - | 2 | 292 |