Hamburger Menu Button
Thermo Fisher Scientific Logo
登录
没有账号?
建立账号
注册
产品
抗体
Custom DNA Oligos
GeneArt Gene Synthesis Instant Designer
TaqMan Assay and Array Search
PCR 设备和耗材
色谱柱和纯化柱
实验室离心机
分子生物学试剂和试剂盒
移液器和移液器吸头
查看所有产品分类
应用和技术
色谱
细胞转染和培养
实时荧光定量PCR
质谱
实验室塑料耗材及用品
电子显微镜
蛋白质生物学
PCR
RNAi
环境
查看所有应用和技术
服务
定制服务
企业级实验室信息化服务
企业服务
金融&租赁服务
仪器服务
OEM和商业供应
培训服务
药物开发生产和临床实验服务
查看所有帮助和支持
帮助和支持
学习中心
联系我们
更改国家或地区
查看所有帮助和支持
联系我们
货号直购
订单查询
查找文件&证书
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
搜索全部或下列指定分类
Search
Search button
Search button
Close
联系我们
货号直购
登录
登录
没有账号?
建立账号
注册
我的账号
订单
定制产品及项目
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OLFR464 SIRNA Assays ›
Gene Symbol:
Olfr464
Gene Name:
olfactory receptor 464
Gene Aliases:
MOR240-2
Chromosome Location:
Chr.11: 87913920 - 87915003 on Build GRCm38
UniGene ID:
Mm.271337
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s111886
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
添加到购物车
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Olfr464
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_146412.2
NP_666524.2
1
869
GenBank mRNA
BC120701.1
-
1
870
40129624
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-lhtlm:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0