Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146515.2 | NP_666726.1 | 1 | 126 | ||
GenBank mRNA | BC106777.1 | - | 1 | 126 | 40040271 | |
BC106778.1 | - | 1 | 126 | 40040272 |