Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197097.2 | NP_001184026.2 | 4, 5 | 798 | ||
NM_001197098.1 | NP_001184027.1 | 3, 4 | 440 | |||
NM_002771.3 | NP_002762.2 | 3, 4 | 461 | |||
NM_007343.3 | NP_031369.2 | 3, 4 | 670 | |||
XM_011517965.1 | XP_011516267.1 | 4, 5 | 721 | |||
GenBank mRNA | AB298285.1 | - | 3, 4 | 670 | ||
AB298286.1 | - | 3, 4 | 440 | |||
BC030238.1 | - | 3, 4 | 460 | |||
BC069476.1 | - | 3, 4 | 461 | 7262223 | ||
BC069494.1 | - | 3, 4 | 461 | 7262235 | ||
D45417.1 | - | 3, 4 | 448 | |||
X15505.1 | - | 3, 4 | 461 | |||
X71345.1 | - | 3, 4 | 488 | |||
X72781.1 | - | 3, 4 | 491 |