Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146948.1 | NP_667159.1 | 1 | 586 | ||
GenBank mRNA | BC120521.1 | - | 1 | 612 | 40129444 | |
BC120523.1 | - | 1 | 612 | 40129446 |