Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191037.1 | NP_001177966.1 | 5 | 592 | ||
NM_002807.3 | NP_002798.2 | 5 | 592 | |||
NR_034059.1 | 4 | 422 | ||||
XM_017004517.1 | XP_016860006.1 | 5 | 592 | |||
GenBank mRNA | AK307415.1 | - | 4 | 419 | ||
AK307513.1 | - | 3 | 294 | |||
AK312528.1 | - | 5 | 549 | |||
BC001053.2 | - | 5 | 549 | |||
BC014013.2 | - | 5 | 561 | |||
BC020223.1 | - | 5 | 539 | |||
BC032445.1 | - | 5 | 564 | |||
BC039845.1 | - | 5 | 515 | |||
BC047897.1 | - | 5 | 514 | |||
BC064398.1 | - | 5 | 563 | |||
BC073833.1 | - | 5 | 516 | |||
BC094720.1 | - | 5 | 578 | 4826340 | ||
BC112344.1 | - | 5 | 553 | 40008454 | ||
BC114434.1 | - | 5 | 553 | 40008444 | ||
D44466.1 | - | 5 | 510 |