Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278708.1 | NP_001265637.1 | 15 | 1874 | ||
NM_001278709.1 | NP_001265638.1 | 15 | 1863 | |||
NM_002808.4 | NP_002799.3 | 17 | 2295 | |||
GenBank mRNA | AB209843.1 | - | 17 | 2141 | ||
AK095245.1 | - | 17 | 2686 | |||
AK297256.1 | - | 18 | 2126 | |||
AK301759.1 | - | 15 | 1876 | |||
AK302177.1 | - | 15 | 1863 | |||
AK315474.1 | - | 17 | 2154 | |||
BC002368.2 | - | 17 | 2127 | 2822191 | ||
BC002997.1 | - | 17 | 2148 | 2905429 | ||
BC007897.2 | - | 17 | 2126 | 4131391 | ||
BT009736.1 | - | 17 | 2126 | |||
D78151.1 | - | 17 | 2159 | |||
U12596.1 | - | 17 | 2064 | |||
U18247.1 | - | 13 | 1517 | |||
X86446.1 | - | 17 | 2099 |