Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170597.4 | NP_733485.1 | 2 | 741 | ||
GenBank mRNA | AB046110.1 | - | 2 | 1059 | ||
AK034501.1 | - | 2 | 504 | |||
AK043871.1 | - | 2 | 506 | |||
AK044192.1 | - | 2 | 741 | |||
AK046245.1 | - | 2 | 528 | |||
AK048589.1 | - | 2 | 535 | |||
AK134713.1 | - | 2 | 512 | |||
AK134861.1 | - | 2 | 802 | |||
BC034115.1 | - | 2 | 429 | |||
BC116790.1 | - | 2 | 634 | 40126109 | ||
BC116792.1 | - | 2 | 634 | 40126111 |