Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170756.2 | NP_739562.1 | 3 | 1232 | ||
GenBank mRNA | AK051741.1 | - | 3 | 1178 | ||
AK122364.1 | - | 1 | 1874 | |||
AK144826.1 | - | 3 | 1204 | |||
AK156188.1 | - | 3 | 1233 | |||
AK162743.1 | - | 3 | 1232 | |||
AK170356.1 | - | 3 | 1233 | |||
AK171240.1 | - | 3 | 1206 | |||
AK171664.1 | - | 3 | 1210 | |||
AK171852.1 | - | 3 | 1222 | |||
AK189566.1 | - | 1 | 15 | |||
BC034597.1 | - | 3 | 1221 | 5363513 |