Search
Search

Cyb5r4| Gene Symbol: Cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024195.2 | NP_077157.2 | 13 | 1380 | ||
| GenBank mRNA | AF338818.1 | - | 12 | 1128 | ||
| AK012855.1 | - | 12 | 1018 | |||
| AK034878.1 | - | 13 | 1324 | |||
| AK078682.1 | - | 6 | 586 | |||
| AK090159.1 | - | 13 | 1382 | |||
| AK135261.1 | - | 12 | 1031 | |||
| AK157312.1 | - | 12 | 1384 | |||
| AK167436.1 | - | 12 | 1078 | |||
| AK169937.1 | - | 12 | 1378 | |||
| AK169995.1 | - | 12 | 1375 | |||
| AK172252.1 | - | 12 | 1333 | |||
| AK207052.1 | - | 1 | 35 | |||
| AY321368.1 | - | 11 | 1152 | |||
| AY321369.1 | - | 10 | 999 | |||
| BC002170.1 | - | 6 | 580 | |||
| BC025438.1 | - | 13 | 1391 | 5038563 | ||