Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010661.2 | NP_034791.2 | 1 | 416 | ||
GenBank mRNA | AK053362.1 | - | 1 | 417 | ||
AK143182.1 | - | 1 | 415 | |||
BC131911.1 | - | 1 | 412 | 40130188 | ||
CK627700.1 | - | 1 | 402 | |||
U02880.1 | - | 1 | 413 |