Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172951.3 | NP_766539.2 | 14, 15 | 1427 | ||
GenBank mRNA | AK008673.1 | - | 14, 15 | 1337 | ||
AK035310.1 | - | 14, 15 | 1381 | |||
AK087560.1 | - | 14, 15 | 1377 | |||
AK137345.1 | - | 14, 15 | 1387 | |||
BC058204.1 | - | 13, 14 | 1189 | |||
BC138956.1 | - | 14, 15 | 1341 | 8861978 | ||
BC145476.1 | - | 14, 15 | 1336 |