Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172958.3 | NP_766546.1 | 15 | 1630 | ||
GenBank mRNA | AK030139.1 | - | 15 | 1697 | ||
AK050099.1 | - | 15 | 1623 | |||
AK050236.1 | - | 15 | 1593 | |||
AK076161.1 | - | 7 | 710 | |||
AK122539.1 | - | 15 | 1688 | |||
BC054352.1 | - | 15 | 1630 | 3661020 |