Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270482.1 | NP_001257411.1 | 13 | 1273 | ||
NM_002815.3 | NP_002806.2 | 13 | 1273 | |||
GenBank mRNA | AB003102.1 | - | 13 | 1223 | ||
AF001212.1 | - | 13 | 1252 | |||
AK093602.1 | - | 3 | 1937 | |||
AK223196.1 | - | 13 | 1229 | |||
AK290602.1 | - | 13 | 1262 | |||
BC000437.2 | - | 13 | 1220 | 2820583 | ||
BC004430.1 | - | 13 | 1220 | 2820583 |