Search
Search

Atp5h| Gene Symbol: Atp5h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027862.1 | NP_082138.1 | 4 | 275 | ||
| GenBank mRNA | AF354051.1 | - | 3 | 268 | ||
| AK002376.1 | - | 4 | 275 | |||
| AK088617.1 | - | 3 | 270 | |||
| BC016547.1 | - | 4 | 273 | 2631309 | ||
| BC081431.1 | - | 4 | 274 | 6408035 | ||