Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166033.1 | NP_001159505.1 | 3 | 879 | ||
NM_172977.3 | NP_766565.2 | 2 | 773 | |||
GenBank mRNA | AK030351.1 | - | 2 | 700 | ||
AK141269.1 | - | 2 | 754 | |||
AK146401.1 | - | 3 | 835 | |||
AK146403.1 | - | 3 | 835 | |||
AK150650.1 | - | 2 | 755 | |||
AK151179.1 | - | 2 | 755 | |||
AK153102.1 | - | 2 | 755 | |||
BC061476.1 | - | 2 | 706 | |||
BC095998.1 | - | 2 | 712 | 30736671 | ||
BC117919.1 | - | 2 | 685 | 40097451 | ||
BC117920.1 | - | 2 | 683 | 40097456 |