Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147153.3 | NP_671495.1 | 10 | 1003 | ||
NM_178851.3 | NP_849182.1 | 9 | 970 | |||
GenBank mRNA | AB028844.1 | - | 9 | 950 | ||
AF281050.1 | - | 10 | 980 | |||
AF281051.1 | - | 9 | 947 | |||
AK038824.1 | - | 9 | 959 | |||
AK042253.1 | - | 9 | 959 | |||
AK084743.1 | - | 9 | 970 | |||
AK129210.1 | - | 9 | 965 | |||
AK140041.1 | - | 10 | 1008 | |||
AK141424.1 | - | 9 | 953 | |||
AK154103.1 | - | 9 | 961 | |||
AK159889.1 | - | 9 | 963 | |||
AK160109.1 | - | 9 | 963 | |||
AK170831.1 | - | 10 | 1030 | |||
AK171008.1 | - | 9 | 959 | |||
BC007479.1 | - | 9 | 921 | 3492875 |