Search
Search

Plch1| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177732.1 | NP_001171203.1 | 24 | 4898 | ||
| NM_001177733.1 | NP_001171204.1 | 24 | 4893 | |||
| NM_183191.3 | NP_899014.2 | 23 | 4870 | |||
| GenBank mRNA | AK173095.1 | - | 14 | 3693 | ||
| AY691172.1 | - | 24 | 4898 | |||
| AY691173.1 | - | 24 | 4893 | |||
| AY691174.1 | - | 23 | 4870 | |||
| BC042549.1 | - | 11 | 3162 | |||
| BC052372.1 | - | 11 | 3162 | |||
| BC055005.1 | - | 22 | 4884 | 6491217 | ||
| BC057610.1 | - | 3 | 2460 | |||